Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKθ Mouse, Unlabeled, Clone: 24, BD

Catalog No. BDB610931 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610931 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610931 Supplier BD Biosciences Supplier No. 610931

Mouse Monoclonal Antibody

The protein kinase C pathway is a major signal transduction system that is activated upon stimulation of transmembrane receptors by hormones, neurotransmitters, and growth factors. Key mediators in this pathway are increased intracellular free Ca2+ levels and formation of diacylglycerol (DAG). DGKθ (diacylglycerol kinase θ) restricts PKC activation through the phosphorylation of DAG molecules that contain an unsaturated fatty acid at the sn-2 position to produce phosphatidic acid (PA). DGKθ contains several regions that are found in signaling molecules where they function in lipid-protein and protein-protein interactions. A C-terminal catalytic domain, three CRDs (cysteine rich domains), a PH domain, and an N-terminal proline/glycine rich domain are structural features of DGKθ. Six potential PKC phosphorylation sites lie between CRD3 and the PH domain. Cell-specific expression differentiate multiple isoforms of DGK. DGKθ is expressed primarily within the cerebellar cortex and hippocampus of the brain, but is also found in the small intestine and liver. The presence of the RA (Ras-associating) domain suggests that DGKθ may mediate activity of the Ras-like small GTP binding proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGKθ
Applications Western Blot
Classification Monoclonal
Clone 24
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human DGKθ aa. 677-883
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.