Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGUOK, Mouse, Clone: 4E5, Abnova™

Catalog No. 89021297 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-297 Supplier Abnova Corporation Supplier No. H00001716M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DGUOK.

In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predominantly by two deoxyribonucleoside kinases, cytosolic deoxycytidine kinase and mitochondrial deoxyguanosine kinase. The protein encoded by this gene is responsible for phosphorylation of purine deoxyribonucleosides in the mitochondrial matrix. In addition, this protein phosphorylates several purine deoxyribonucleoside analogs used in the treatment of lymphoproliferative disorders, and this phosphorylation is critical for the effectiveness of the analogs. Alternative splice variants encoding different protein isoforms have been described for this gene. [provided by RefSeq

Sequence: MAAGRLFLSRLRAPFSSMAKSPLEGVSSSRGLHAGRGPRRLSIEGNIAVGKSTFVKLLTKTYPEWHVATEPVATWQNIQAAGTQKACTAQSLGNLLDMMYREPARWSYTFQTFSFLSRLKVQLEPFPEKLLQARKPVQIFERSVYSDRYIFAKNLFENGSLSDIEWHIYQDWHSFLLWEFASRITLHGFIYLQASPQVCLKRLYQRAREEEKGIELAYLEQLHGQHEAWLIHKTTKLHFEALMNIPVLVLDVNDDFSEEVTKQEDLMREVNTFVKNL

Specifications

Antigen DGUOK
Applications ELISA
Classification Monoclonal
Clone 4E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DGUOK.
Formulation PBS with no preservative; pH 7.4
Gene DGUOK
Gene Accession No. BC015757
Gene Alias dGK
Gene Symbols DGUOK
Host Species Mouse
Immunogen DGUOK (AAH15757, 1 a.a. ∼ 277 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1716
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.