Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHCR24, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002937 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-937 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-937 Supplier Abnova Corporation Supplier No. H00001718D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human DHCR24 protein.

This gene encodes a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. The protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in this gene have been associated with desmosterolosis. Also, reduced expression of the gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells. [provided by RefSeq

Sequence: MEPAVSLAVCALLFLLWVRLKGLEFVLIHQRWVFVCLFLLPLSLIFDIYYYVRAWVVFKLSSAPRLHEQRVRDIQKQVREWKEQGSKTFMCTGRPGWLTVSLRVGKYKKTHKNIMINLMDILEVDTKKQIVRVEPLVTMGQVTALLTSIGWTLPVLPELDDLTVGGLIMGTGIESSSHKYGLFQHICTAYELVLADGSFVRCTPSENSDLFYAVPWSCGTLGFLVAAEIRIIPAKKYVKLRFEPVRGLEAICAKFTHESQRQENHFVEGLLYSLDEAVIMTGVMTDEAEPSKLNSIGNYYKPWFFKHVENYLKTNREGLEYIPLRHYYHRHTRSIFWELQDIIPFGNNPIFRYLFGWMVPPKISLLKLTQGETLRKLYEQHHVVQDMLVPMKCLQQALHTFQNDIHVYPIWLCPFILPSQPGLVHPKGNEAELYIDIGAYGEPRVKHFEARSCMRQLEKFVRSVHGFQMLYADCYMNREEFWEMFDGSLYHKLREKLGCQDAFPEVYDKICKAARH

Specifications

Antigen DHCR24
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human DHCR24 protein.
Formulation PBS with no preservative; pH 7.4
Gene DHCR24
Gene Accession No. NM_014762.2
Gene Alias KIAA0018/Nbla03646/SELADIN1/seladin-1
Gene Symbols DHCR24
Host Species Rabbit
Immunogen DHCR24 (NP_055577.1, 1 a.a. ∼ 516 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1718
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.