Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHCR24 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512457
Description
This antibody is predicted to react with bovine, chicken, mouse and rat based on sequence homology.
DHCR24 is a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. This protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in DHCR24 gene have been associated with desmosterolosis. Also, reduced expression of DHCR24 gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells.
Specifications
DHCR24 | |
Polyclonal | |
DHCR24 | |
Diminuto/dwarf1 homolog; Seladin-1; 3-beta-hydroxysterol delta-24-reductase; 24-dehydrocholesterol reductase; DHCR24; KIAA0018 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
1718 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q15392 | |
DHCR24 | |
KLH conjugated synthetic peptide between 218-246 amino acids from the central region of human DHCR24 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction