Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dehydrogenase/reductase (SDR family) member 4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-082 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-082 Supplier Abnova Corporation Supplier No. H00010901B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DHRS4 protein.

Sequence: MHKAGLLGLCARAWNSVRMASSGMTRRDPLANKVALVTASTDGIGFAIARRLAQDGAHVVVSSRKQQNVDQAVATLQGEGLSVTGTVCHVGKAEDRERLVATAVKLHGGIDILVSNAAVNPFFGSIMDVTEEVWDKTLDINVKAPALMTKAVVPEMEKRGGGSVVIVSSIAAFSPSPGFSPYNVSKTALLGLTKTLAIELAPRNIRVNCLAPGLIKTSFSRMLWMDKEKEESMKETLRIRRLGEPEDCAGIVSFLCSEDASYITGETVVVGGGTPSRL

Specifications

Antigen dehydrogenase/reductase (SDR family) member 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DHRS4 protein.
Formulation No additive
Gene DHRS4
Gene Accession No. NM_021004.2
Gene Alias DHRS4L2/FLJ11008/NRDR/SCAD-SRL/SDR-SRL/SDR25C1/humNRDR
Gene Symbols DHRS4
Host Species Mouse
Immunogen DHRS4 (NP_066284.2, 1 a.a. ∼ 278 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10901
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.