Learn More
DHRS7, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DHRS7 peptide using ARM Technology.
Supplier: Abnova Corporation H00051635K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DHRS7 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DHRS7 | |
DHRS7 | |
A synthetic peptide of human DHRS7 | |
RUO | |
51635 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
dehydrogenase/reductase (SDR family) member 7 | |
DHRS7 | |
CGI-86, SDR34C1, retDSR4, retSDR4 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.