Learn More
DHX8, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DHX8 peptide using ARM Technology.
Supplier: Abnova Corporation H00001659K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is highly homologous to yeast Prp22. This protein facilitates nuclear export of spliced mRNA by releasing the RNA from the spliceosome. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DHX8 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DHX8 | |
DHX8 | |
A synthetic peptide of human DHX8 | |
RUO | |
1659 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
DEAH (Asp-Glu-Ala-His) box polypeptide 8 | |
DHX8 | |
DDX8, HRH1, PRP22, PRPF22 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.