Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diglycyl Lysine Mouse anti-Rat, All, Human, Mouse, Clone: GX41, MilliporeSigma™

Catalog No. MABS27MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS27MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS27MI Supplier MilliporeSigma Supplier No. MABS27
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Diglycyl Lysine (Clone GX41) Antibody detects level of Diglycyl Lysine (Clone GX41) and has been published and validated for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Diglycyl Lysine
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GX41
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols Diglycyl Lysine
Host Species Mouse
Immunogen Linear peptide corresponding to Diglycyl Lysine.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species All species, Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.