Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dihydropyridine-sensitive Calcium Channel α 1 Subunit Mouse anti-Human, Mouse, Rat, Rabbit, Clone: mAB 1a, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB427
Description
Anti-Dihydropyridine-sensitive Calcium Channel α 1 Subunit Antibody, clone 1a is an antibody against Dihydropyridine-sensitive Calcium Channel α 1 Subunit for use in flow cytometry, immunoprecipitation and western blotting.
Specifications
Dihydropyridine-sensitive Calcium Channel α 1 Subunit | |
Monoclonal | |
Unconjugated | |
NM_000069.2 | |
Mouse | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
mAB 1a | |
Ascites. | |
CACNA1S, CACH1, CCHL1A3, hypoPP, MHS5, CACNL1A3, Cav1.1, HOKPP, CACN1, HypoPP | |
100 μL | |
Human, Mouse, Rabbit, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction