Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Arginine Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07414MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-414-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-414-MI Supplier MilliporeSigma Supplier No. 07414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect dimethyl-Arginine with Anti-dimethyl-Arginine Antibody, asymmetric (ASYM24) (Rabbit Polyclonal Antibody), that has been shown to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Arginine, asymmetric (ASYM24)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum diluted in storage buffer (PBS, 0.035% sodium azide and 30% glycerol). Store at −20°C.
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing asymmetric dimethyl-arginine-glycine repeats (KGR*GR*GR*GR*GPPPPPR*GR*GR*GR*G in which R* denotes asymmetric dimethylation of arginine).
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.