Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Arginine Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07412MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-412-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-412-MI Supplier MilliporeSigma Supplier No. 07412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-dimethyl-Arginine Antibody, symmetric (SYM10) is a high quality Rabbit Polyclonal Antibody for the detection of dimethyl-Arginine and has been validated in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Arginine, symmetric (SYM10)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Whole rabbit antiserum containing 0.05% sodium azide and 30% glycerol. Liquid at −20°C.
Host Species Rabbit
Immunogen peptide containing four symmetrical dimethyl-arginine-glycine repeats
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.