Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl-Arginine Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07413
Description
Detect dimethyl-Arginine with Anti-dimethyl-Arginine Antibody, symmetric (SYM11) (Rabbit Polyclonal Antibody), that has been shown to work in western blotting.
Specifications
| dimethyl-Arginine, symmetric (SYM11) | |
| Polyclonal | |
| 0.1M Tris-glycine, pH 7.4, 0.15M NaCl,0.05% sodium azide before the addition of glycerol to 30%. | |
| peptide corresponding to amino acids 99-119 of Small nuclear ribonucleoprotein SmD3. | |
| 200 μg | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein A purfied | |
| Primary | |
| −20°C for 24 months | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction