Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Histone H3 (Arg2) Rabbit anti-Chicken, Human, Polyclonal, MilliporeSigma™

Catalog No. 07585MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-585-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-585-MI Supplier MilliporeSigma Supplier No. 07585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-dimethyl-Histone H3 (Arg2) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl-Histone H3 (Arg2) also known as H3R2me2, Histone H3 (di methyl R2) and has been validated in western blot, PIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Histone H3 (Arg2)
Applications Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation rabbit antiserum containing 0.05% sodium azide, before the addition of glycerol to 30%
Gene Symbols H3F3A; MGC87783; H3.3A; MGC87782; H3F3; H3.3B; H3F3B
Host Species Rabbit
Immunogen peptide containing the sequence AdimRT, corresponding to dimethyl-arginine 2 of human histone H3
Purification Method Serum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Chicken, Human
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.