Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl Histone H3 (Arg2), symmetric Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE460
Description
This Anti-dimethyl Histone H3 (Arg2) antibody, symmetric is validated for use in western blotting, dot blot, immunohistochemistry and Chimmunoprecipitation for the detection of dimethyl Histone H3 (Arg2).
Specifications
dimethyl Histone H3 (Arg2), Symmetric | |
Polyclonal | |
Rabbit polyclonal serum containing 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Human | |
Serum |
ChIP Assay, Dot Blot, Immunohistochemistry, Western Blot | |
Unconjugated | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ | |
Linear peptide corresponding to human Histone H3 symmetrically dimethylated at Arg2. | |
100 μL | |
NP_003520 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction