Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Histone H3 (Lys36) Rabbit anti-Mouse, Human, Chicken, Drosophila, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Catalog No. 07274MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-274-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-274-MI Supplier MilliporeSigma Supplier No. 07274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-dimethyl-Histone H3 (Lys36) Antibody (Rabbit Polyclonal Antibody) validated in western blot, DB to detect dimethyl-Histone H3 (Lys36) also known as H3K36me2, Histone H3 (di methyl K36).
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Histone H3 (Lys36)
Applications ChIP Assay, Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG immunodepleted antiserum in buffer containing 0.05% sodium azide before the addition of glycerol to 30%. Liquid at −20°C. Serum was passed over immunoaffinity column containing the non-modified peptide.
Gene Symbols HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 30-38 (CPATGGV(dimethyl-K)KP) of Histone H3 with a N-terminal C added for conjugation. The immunizing sequence is conserved from Tetrahymena to human.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Chicken, Drosophila, Human, Mouse, S. cerevisiae, Vertebrates, Yeast
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.