Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Histone H3 (Lys4) Rabbit anti-Human, Chicken, Clone: AW30, MilliporeSigma™

Catalog No. 04790MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-790-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-790-MI Supplier MilliporeSigma Supplier No. 04790
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30 is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4) and has been validated in western blot, Chimmunoprecipitation, ChIP-seq, DB, PIA, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Histone H3 (Lys4)
Applications ChIP Assay, Dot Blot, Inhibition Assays, Western Blot
Classification Monoclonal
Clone AW30
Conjugate Unconjugated
Formulation Cultured supernantant in 0.05% sodium azide
Gene Accession No. NM_003493
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence ...Tme2KQT... in which me2K corresponds to dimethyl-lysine at residue 4 of human histone H3.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) HIST3H3, H3FT, MGC126886, H3t, MGC126888, H3T, H3/g, H3.4, H3/t
Target Species Chicken, Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.