Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Histone H3 (Lys4) Mouse anti-Human, Vertebrates, Clone: CMA303, MilliporeSigma™

Mouse Monoclonal Antibody

$144.24 - $657.06

Specifications

Antigen Dimethyl Histone H3 (Lys4)
Clone CMA303
Applications ChIP Assay, Dot Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05-133-8SMI
View Documents
MilliporeSigma
051338S
10 μg
Each for $144.24
Only null left
Add to Cart
 
05-133-8MI
View Documents
MilliporeSigma
051338
100 μg
Each for $657.06
Only null left
Add to Cart
 
Description

Description

Anti-Dimethyl Histone H3 (Lys4) Antibody, clone CMA303 is a mouse monoclonal antibody for detection of Dimethyl Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4). Demonstrated performance in Chimmunoprecipitation, DB, ELISA, immunocytochemistry, immunoprecipitation, western blot, Mplex.
Specifications

Specifications

Dimethyl Histone H3 (Lys4)
ChIP Assay, Dot Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.
H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
IgG1 κ
Protein G purfied
CMA303
Monoclonal
Purified
Human, Vertebrates
NM_003493
Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, dimethylated on Lys4, conjugated to KLH.
Primary
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.