Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-dimethyl-Histone H3 (Lys4), Clone: RR302,

Catalog No. 05684MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-684-MI 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-684-MI Supplier MilliporeSigma™ Supplier No. 05684

Specifications

Antigen dimethyl-Histone H3 (Lys4)
Applications Western Blot
Classification Monoclonal
Clone RR302
Conjugate Unconjugated
Formulation PBS containing 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_003493
Gene Symbols H3F3A, H3F3, MGC87782, H3.3B, MGC87783, H3F3B, H3.3A
Host Species Mouse
Immunogen peptide (RTdimKQ) corresponding to dimethyl lysine at position 4 of human Histone H3
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Primary or Secondary Primary
Target Species Chicken, Human, Vertebrates
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.