Learn More
Dimethyl-Histone H3 (Lys4) Rabbit anti-Human, Tetrahymena, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | dimethyl-Histone H3 (Lys4) |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Immunofluorescence, Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal antiserum containing 0.05% sodium azide. |
| Gene Symbols | HIST3H3; H3FT; H3/t; H3t; MGC126888; H3/g; H3.4; H3T; MGC126886 |
| Host Species | Rabbit |
| Immunogen | KLH conjugated synthetic peptide (ARTMe2KQTAR-GC) corresponding to amino acids 1-8 of human Histone H3. |
| Purification Method | Unpurified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.