Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Histone H3 (Lys4) Rabbit anti-Human, Mouse, Rat, Chicken, Polyclonal, MilliporeSigma™

Catalog No. ABE250MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE250MI 100 μL
ABE250SMI 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE250MI Supplier MilliporeSigma Supplier No. ABE250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-dimethyl Histone H3 (Lys4) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 dimethylated at lysine 4. This purified polyclonal antibody, also known as H3K4me2, has dot blot (DB) proven specificity and has been validated in western blot, immunocytochemistry, Chimmunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl Histone H3 (Lys4)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding Histone H3 methylated at Lys4.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Target Species Chicken, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.