Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl-Histone H3 (Lys79) Rabbit anti-Human, Vertebrates, Clone: NL59, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04835
Description
Anti-dimethyl-Histone H3 (Lys79) Antibody, clone NL59 is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys79) also known as H3K79me2, Histone H3 (di methyl K79) and has been validated in Chimmunoprecipitation, western blot, DB, Mplex.
Specifications
dimethyl-Histone H3 (Lys79) | |
Monoclonal | |
Unconjugated | |
NM_003493 | |
KLH-conjugated synthetic peptide containing the sequence DFme2KTD corresponding to dimethyl-lysine at position 79 of histone H3. The immunizing sequence is identical in human, mouse, rat and yeast. A C-terminal cysteine was added to facilitat | |
Primary | |
Human, Vertebrates | |
Supernatant |
Multiplex, Dot Blot, Immunoassay, Western Blot | |
NL59 | |
Culture supernantant in 0.05% sodium azide | |
Rabbit | |
100 μL | |
H3F3A, H3F3B | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction