Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Histone H3 (Lys79) Rabbit anti-Human, Vertebrates, Clone: NL59, MilliporeSigma™

Catalog No. 04835SMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-835-SMI 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-835-SMI Supplier MilliporeSigma Supplier No. 04835S
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-dimethyl-Histone H3 (Lys79) Antibody, clone NL59 is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys79) also known as H3K79me2, Histone H3 (di methyl K79) and has been validated in Chimmunoprecipitation, western blot, DB, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Histone H3 (Lys79)
Applications Multiplex, Dot Blot, Immunoassay, Western Blot
Classification Monoclonal
Clone NL59
Conjugate Unconjugated
Formulation Culture supernantant in 0.05% sodium azide
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide containing the sequence DFme2KTD corresponding to dimethyl-lysine at position 79 of histone H3. The immunizing sequence is identical in human, mouse, rat and yeast. A C-terminal cysteine was added to facilitat
Quantity 10 μL
Primary or Secondary Primary
Gene ID (Entrez) H3F3A, H3F3B
Target Species Human, Vertebrates
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.