Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-Histone H3 (Lys9)-Alexa Fluor™488 Rabbit anti-Human, AlexaFluor™488, Polyclonal, MilliporeSigma™

Catalog No. FCABS301A4 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCABS301A4 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCABS301A4 Supplier MilliporeSigma Supplier No. FCABS301A4
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Milli-Mark™ Anti-dimethyl-Histone H3 (Lys9)-Alexa Fluor™488 Antibody detects level of dimethyl-Histone H3 (Lys9)-Alexa Fluor™488 and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl-Histone H3 (Lys9)
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal IgG conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15mg/mL BSA
Gene Symbols HIST3H3; H3FT; H3/t; H3t; MGC126888; H3/g; H3.4; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated, 2X-branched synthetic peptide corresponding to dimethyl-lysine at residue 9 of human Histone H3.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Target Species Human
Content And Storage 2-8°C protected from light for up to 6 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.