Learn More
Dimethyl-Histone H3 (Lys9) Rabbit anti-Human, Chicken, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | dimethyl-Histone H3 (Lys9) |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal IgG in buffer containing 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 5mg/mL BSA, 0.05% sodium azide before the addition of glycerol to 30%. |
| Gene Symbols | HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated synthetic peptide corresponding to amino acids 6-13 (TAR[dimethyl-K]STGG-C) of Histone H3 with C added to the Cterminus for conjugation. The immunizing sequence is conserved from Tetrahymena to human. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.