Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Histone H4 (Lys20) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071584MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-158-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-158-4MI Supplier MilliporeSigma Supplier No. 071584

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Anti-dimethyl Histone H4 (Lys20) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl Histone H4 (Lys20) also known as H4K20me2, Histone H4 (di methyl K20) and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl Histone H4 (Lys20)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence (AKRHR[dimethyl-K]VLRD-C), corresponding to the sequence surrounding acetylated lysine 20 of human histone H4.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_175054
Target Species Human
Content And Storage 2-8°C for 12 months For maximum recovery of product centrifuge the vial prior to removing the cap.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.