Learn More
DIP2 disco-interacting protein 2 homolog A (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human DIP2A protein.
Supplier: Abnova Corporation H00023181D01P
Description
The protein encoded by this gene may be involved in axon patterning in the central nervous system. This gene is not highly expressed. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MADRGCPLEAAPLPAEVRESLAELELELSEGDITQKGYEKKRAKLLARYIPLIQGIDPSLQAENRIPGPSQTTAAAPKQQKSRPTASRDERFRSDVHTEAVQAALAKYKERKMPMPSKRRSVLVHSSVETYTPPDTSSASEDEGSLRRPGRLTSTPLQSHSSVEPWLDRVIQGSSTSSSASSTSSHPGGRPTTAPSAAATPGAAATTALAGLEAHTHIDLHSAPPDVTTGLVEHSYFERPQVASVRSVPRGCSGSMLETADGVPVNSRVSSKIQQLLNTLKRPKRPPLKEFFVDDFEELLEVQQPDPNQPKPEGSETSVLRGEPLTAGVPRPPSLLATLQRWGTTQPKSPCLTALDTTGKAVYTLTYGKLWSRSLKLAYTLLNKLTSKNEPLLKPGDRVALVFPNSDPVMFMVAFYGCLLAELVPVPIEVPLTRKDAGSQQVGFLLGSCGVFLALTTDACQKGLPKAQTGEVAAFKGWPPLSWLVIDGKHLAKPPKDWHPLAQDTGTGTAYIEYKTSKEGSTVGVTVSHASLLAQCRALTQACGYSEAETLTNVLDFKRDAGLWHGVLTSVMNRMHVVSVPYALMKANPLSWIQKVCFYKARAALVKSRDMHWSLLAQRGQRDVSLSSLRMLIVADGANPWSISSCDAFLNVFQSRGLRPEVICPCASSPEALTVAIRRPPDLGGPPPRKAVLSMNGLSYGVIRVDTEEKLSVLTVQDVGQVMPGANVCVVKLEGTPYLCKTDEVGEICVSSSATGTAYYGLLGITKNVFEAVPVTTGGAPIFDRPFTRTGLLGFIGPDNLVFIVGKLDGLMVTGVRRHNADDVVATALAVEPMKFVYRGRIAVFSVTVLHDDRIVLVAEQRPDASEEDSFQWMSRVLQVGAPARPMVRSpecifications
DIP2 disco-interacting protein 2 homolog A (Drosophila) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human DIP2A protein. | |
DIP2A | |
C21orf106/DIP2 | |
Rabbit | |
Affinity Purified | |
RUO | |
23181 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_206889.1 | |
DIP2A | |
DIP2A (NP_996772.1, 1 a.a. ∼ 889 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.