Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disabled-2/p96 Mouse, Unlabeled, Clone: 52, BD

Catalog No. BDB610464 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610464 50 μg
BDB610465 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610464 Supplier BD Biosciences Supplier No. 610464
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSF-1 is a growth factor that stimulates the growth and differentiation of immature lymphocytes and is required for the survival of mononuclear phagocytes. Binding of CSF-1 induces dimerization and autophosphorylation of its receptor. This results in the activation of several signal transduction pathways. A unique 96 kDa protein is a component in the CSF-1 signal transduction cascade. p96 is phosphorylated on serine following mitogenic stimulation of a mouse macrophage cell line. p96 contains three potential C-terminal ERK kinase phosphorylation sites, as well as several proline-rich sequences that are potential binding sites for SH3-containing proteins. Structural similarities have been found between p96 and Dab, a product of the Drosophila disabled gene, and p96 was also identified as Disabled-2 (Dab-2) and as differentially expressed in ovarian β receptor to the Smad family of transcription factors. Thus, Dab-2/p96 is an important adaptor molecule in growth factor signaling pathways.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Disabled-2/p96
Applications Western Blot
Classification Monoclonal
Clone 52
Concentration 250μg/mL
Conjugate Unconjugated
Description Disabled-2
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.