Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disialoganglioside GD2 Mouse anti-Rat, Mouse, AlexaFluor™488, Clone: 14G2a, MilliporeSigma™

Catalog No. MAB2052A4MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2052A4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2052A4MI Supplier MilliporeSigma Supplier No. MAB2052A4

Mouse Monoclonal Antibody

Detect Disialoganglioside GD2 using this Anti-Disialoganglioside GD2 Antibody, clone 14G2a, AlexaFluor™488 Conj. validated for use in immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Disialoganglioside GD2
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 14.2Ga
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG2a conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols GD2
Host Species Mouse
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.