Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disialoganglioside GD2 Mouse anti-Human, Clone: 14G2a, MilliporeSigma™

Catalog No. MAB2052MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2052MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2052MI Supplier MilliporeSigma Supplier No. MAB2052
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Disialoganglioside GD2 Antibody, clone 14G2a detects level of Disialoganglioside GD2 and has been published and validated for use in flow cytometry, immunofluoresence, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Disialoganglioside GD2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 14.2Ga
Conjugate Unconjugated
Formulation Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing 0.1% NaN3.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.