Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disialosyl LEA Antigen Mouse anti-Human, Clone: FH7, MilliporeSigma™

Catalog No. MAB10210MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10210MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10210MI Supplier MilliporeSigma Supplier No. MAB10210
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Disialosyl LEA Antigen Antibody, clone FH7 detects level of Disialosyl LEA Antigen and has been published and validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Disialosyl LEA Antigen
Applications Immunofluorescence
Classification Monoclonal
Clone FH7
Conjugate Unconjugated
Formulation Liquid ascites fluid
Host Species Mouse
Immunogen Purified Disialosyl Lea antigen
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for one year
Form Ascites
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.