Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLC-1 Mouse anti-Human, Unlabeled, Clone: 3, BD

Catalog No. BDB612020 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612020 50 μg
BDB612021 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612020 Supplier BD Biosciences Supplier No. 612020
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the Ras superfamily of GTPases are distantly related to the heterotrimeric G proteins and shuttle between inactive and active GTP-bound forms. Conversion between these forms is controlled by guanine nucleotide exchange factors (GEFs) and GTPase-activating proteins (GAPs). Rho is a GTPase that regulates cell morphology and motility in response to extracellular signals. p190A, p190B, and p122 RhoGAPs all contain RhoGAP domains and regulate the GTPase activity of Rho. p122 RhoGAP exhibits GAP activity for RhoA, but not Rac1, and binds and activates PLC-δ1. Microinjection of p122 RhoGAP suppresses LPA-induced formation of stress fibers and focal adhesions. Deleted in liver cancer-1 (DLC-1) was identified by its frequent deletion in liver cancer and is a human homologue of rat p122 RhoGAP. DLC-1 is expressed in many hepatocellular carcinoma tissues and cell lines and could be an important tumor suppressor gene. In normal human tissues, DLC-1 is expressed widely with the highest expression in heart and lung. Both p122 RhoGAP and DLC-1 have RhoGAP domains toward their C-terminal regions, which is thought to be important for regulation of Rho GTPase activity in many tissues.

Host Species: Mouse
Clone: 3
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human DLC-1 aa. 47-249

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen DLC-1
Applications Western Blot
Classification Monoclonal
Clone 3
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human DLC-1 aa. 47-249
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.