Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLC1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-972 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-972 Supplier Abnova Corporation Supplier No. H00010395B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DLC1 protein.

This gene is deleted in the primary tumor of hepatocellular carcinoma. It maps to 8p22-p21.3, a region frequently deleted in solid tumors. It is suggested that this gene is a candidate tumor suppressor gene for human liver cancer, as well as for prostate, lung, colorectal, and breast cancers. Alternative splicing at this locus results in several transcript variants encoding different isoforms. [provided by RefSeq

Sequence: MSVAIRKRSWEEHVTHWMGQPFNSDDRNTACHHGLVADSLQASMEKDATLNVDRKEKCVSLPDCCHGSELRDFPGRPMGHLSKDVDENDSHEGEDQFLSLEASTETLVHVSDEDNNADLCLTDDKQVLNTQGQKTSGQHMIQGAGSLEKALPIIQSNQVSSNSWGIAGETELALVKESGERKVTDSISKSLELCNEISLSEIKDAPKVNAVDTLNVKDIAPEKQLLNSAVIAQQRRKPDPPKDENERSTCNVVQDEFLDTPCTNRGLPLLKTDFGSCLLQPPSCPNGMSAENGLEKSGFSQHQNKSPPKVKAEDGMQCLQLKETLATQEPTDNQVRLRKRKEIREDRDRARLDSMVLLIMKLDQLDQDIENALSTSSSPSGTPTNLRRHVPDLESGSESGADTISVNQTRVNLSSDTESTDLPSSTPVANSGTKPKTTAIQGISEKEKAEIEAKEACDWLRATGFPQYAQLYEDFLFPIDISLVKREHDFLDRDAIEALCRRLNTLNKCAVMKLEISPHRKRSDDSDEDEPCAISGKWTFQRDSKRWSRLEEFDVFSPKQDLVPGSPDDSHPKDGPSPGGTLMDLSERQEVSSVRSLSSTGSLPSHAPPSEDAATPRTNSVISVCSSSNLAGNDDSFGSLPSPKELSSFSFSMKGHEKTAKSKTRSLLKRMESLKLKSSHHSKHKAPSKLGLIISGPILQEGMDEEKLKQLNCVEISALNGNRINVPMVRKRSVSNSTQTSSSSSQSETSSAVSTPSPVTRTRSLSACNKRVGMYLEGFDPFNQSTFNNVVEQNFKNRESYPEDTVFYIPEDHKPGTFPKALTNGSFSPSGNNGSVNWRTGSFHGPGHISLRRENSSDSPKELKRRNSSSSMSSRLSIYDNVPGSILYSSSGDLADLENEDIFPELDDILYHVKGMQRIVNQWSEKFSDEGDSDSALDSVSPCPSSPKQIHLDVDNDRTTPSDLDSTGNSLNEPEEPSEIPERRDSGVGASLTRSNRHRLRWHSFQSSHRPSLNSVSLQINCQSVAQMNLLQKYSLLKLTALLEKYTPSNKHGFSWAVPKFMKRIKVPDYKDRSVFGVPLTVNVQRTGQPLPQSIQQAMRYLRNHCLDQVGLFRKSGVKSRIQALRQMNEGAIDCVNYEGQSAYDVADMLKQYFRDLPEPLMTNKLSETFLQIYQYVPKDQRLQAIKAAIMLLPDENREVLQTLLYFLSDVTAAVKENQMTPTNLAVCLAPSLFHLNTLKRENSSPRVMQRKQSLGKPDQKDLNENLAATQGLAHMIAECKKLFQVPEEMSRCRNSYTEQELKPLTLEALGHLGNDDSADYQHFLQDCVDGLFKEVKEKFKGWVSYSTSEQAELSYKKVSEGPPLRLWRSVIEVPAVPEEILKRLLKEQHLWDVDLLDSKVIEILDSQTEIYQYVQNSMAPHPARDYVVLRTWRTNLPKGACALLLTSVDHDRAPVVGVRVNVLLSRYLIEPCGPGKSKLTYMCRVDLRGHMPEWYTKSFGHLCAAEVVKIRDSFSNQNTETKDTKSR

Specifications

Antigen DLC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DLC1 protein.
Formulation No additive
Gene DLC1
Gene Accession No. NM_182643.1
Gene Alias ARHGAP7/FLJ21120/HP/STARD12/p122-RhoGAP
Gene Symbols DLC1
Host Species Mouse
Immunogen 1 a.a. ∽1528 a.a) full-length human protein., DLC1 (NP_872584.1
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10395
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.