Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dlg Mouse, Unlabeled, Clone: 12, BD

Catalog No. BDB610874 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610874 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610874 Supplier BD Biosciences Supplier No. 610874
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The human homologue of the Drosophila discs large tumor suppressor protein (hDlg) is a member of the MAGUKs (membrane associated guanylate kinases) protein family. Members of this family (PSD-95, ZO-1, ZO-2, and human erythroid p55) are involved in cell structure and signaling events. The hDlg protein consists of several domains: three PDZ (PSD-95/Discs large/ZO-1) domains, an SH3 domain, and a guanylate kinase-like domain. However, hDlg contains a proline rich N-terminus region consisting of two SH3 domain binding sites that are not normally found in the MAGUKs family. The PDZ domains mediate the interaction of several proteins, such as Shaker-type K+ channel proteins and the APC tumor suppressor protein. Dlg is a peripheral membrane that associates with the cytoskeleton. The cellular location and binding sites of Dlg suggest a role in structure, signal transduction, and growth regulation. Supporting these probable Dlg functions are reports demonstrating that recessive mutations in Drosophila dlg lead to imaginal disc neoplasia and death. Also, Dlg has been reported to bind p56 [lck] tyrosine kinase and the Kv1.3 channel in human T lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dlg
Applications Western Blot
Classification Monoclonal
Clone 12
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Dlg aa. 5-213
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.