Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

distal-less homeobox 1, Mouse, Clone: 1B2, Abnova™

Catalog No. 89000606 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-606 Supplier Abnova Corporation Supplier No. H00001745M12
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DLX1.

This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein is localized to the nucleus where it may function as a transcriptional regulator of signals from multiple TGF-{beta} superfamily members. The encoded protein may play a role in the control of craniofacial patterning and the differentiation and survival of inhibitory neurons in the forebrain. This gene is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 2. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Sequence: SKFKKLMKQGGAALEGSALANGRALSAGSPPVPPGWNPNSSSGKGSGGNAGSYIPSYTSWYPSAHQEAMQQPQL*

Specifications

Antigen distal-less homeobox 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant DLX1.
Formulation PBS with no preservative; pH 7.4
Gene DLX1
Gene Accession No. NM_178120
Gene Symbols DLX1
Host Species Mouse
Immunogen DLX1 (NP_835221, 181 a.a. ∼ 254 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 1745
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.