Learn More
DLX1, Mouse, Clone: 2H3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DLX1.
Supplier: Abnova Corporation H00001745M01
Description
This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein is localized to the nucleus where it may function as a transcriptional regulator of signals from multiple TGF-{beta} superfamily members. The encoded protein may play a role in the control of craniofacial patterning and the differentiation and survival of inhibitory neurons in the forebrain. This gene is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 2. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq
Sequence: YLALPERAELAASLGLTQTQVKIWFQNKRSKFKKLMKQGGAALEGSALANGRALSAGSPPVPPGWNPNSSSGKGSGGNAGSYIPSYTSWYPSAHQEAMQQPQLMSpecifications
DLX1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_178120 | |
Mouse | |
Affinity chromatography | |
RUO | |
1745 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
2H3 | |
Mouse monoclonal antibody raised against a partial recombinant DLX1. | |
DLX1 | |
DLX1 | |
DLX1 (NP_835221, 152 a.a. ∼ 255 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.