Learn More
DLX2, Mouse, Clone: 2E12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DLX2.
Supplier: Abnova Corporation H00001746M05
Description
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 2. [provided by RefSeq
Sequence: MTGVFDSLVADMHSTQIAASSTYHQHQQPPSGGGAGPGGNSSSSSSLHKPQESPTLPVSTATDSSYYTNQQHPAGGGGGGGSPYAHMGSYQYQASGLNNVPYSAKSSYDLSpecifications
DLX2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004405.3 | |
DLX2 | |
DLX2 (NP_004396.1, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
2E12 | |
Mouse monoclonal antibody raised against a partial recombinant DLX2. | |
DLX2 | |
TES-1/TES1 | |
Mouse | |
Affinity chromatography | |
RUO | |
1746 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.