Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dlx2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5726
Description
Anti-Dlx2 Antibody is an antibody against Dlx2 for use in western blot, immunohistochemistry.
Specifications
Dlx2 | |
Polyclonal | |
Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunohistochemistry, Western Blot | |
Unconjugated | |
DLX2; TES1; TES-1 | |
Synthetic peptide, amino acids 24-39 from human Dlx 2. | |
100 μg | |
NM_004405.3 | |
-70°C for up to 6 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction