Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLX3, Mouse, Clone: 3F7, Abnova™

Catalog No. 89021322 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-322 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-322 Supplier Abnova Corporation Supplier No. H00001747M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DLX3.

Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. Trichodentoosseous syndrome (TDO), an autosomal dominant condition, has been correlated with DLX3 gene mutation. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 17. Mutations in this gene have been associated with the autosomal dominant conditions trichodentoosseous syndrome and amelogenesis imperfecta with taurodontism. [provided by RefSeq

Sequence: MSGSFDRKLSSILTDISSSLSCHAGSKDSPTLPESSVTDLGYYSAPQHDYYSGQPYGQTVNPYTYHHQFNLNGLAGTGAYSPKSEYTYGASYRQYGAYREQPLPAQDPVSVKEEPEAEVRMVNGKPKKVRKPRTIYSSYQLAALQRRFQKAQYLALPERAELAAQLGLTQTQVKIWFQNRRSKFKKLYKNGEVPLEHSPNNSDSMACNSPPSPALWDTSSHSTPAPARSQLPPPLPYSASPSYLDDPTNSWYHAQNLSGPHLQQQPPQPATLHHASPGPPPNPGAVY

Specifications

Antigen DLX3
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DLX3.
Formulation PBS with no preservative; pH 7.4
Gene DLX3
Gene Accession No. BC028970
Gene Alias AI4/TDO
Gene Symbols DLX3
Host Species Mouse
Immunogen DLX3 (AAH28970, 1 a.a. ∼ 287 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1747
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.