Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLX3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | DLX3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224646SS
![]() |
Novus Biologicals
NBP224646SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224646
![]() |
Novus Biologicals
NBP224646 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
DLX3 Polyclonal specifically detects DLX3 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DLX3 | |
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Bovine, Canine, Equine, Primate | |
O60479 | |
1747 | |
A portion of amino acids 220-270 of DLX3 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Growth and Development, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
AI4, distal-less homeo box 3, distal-less homeobox 3, homeobox protein DLX-3, TDO | |
DLX3 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, horse, rat, chimpanzee, rhesus monkey, mouse, dog, 94% homologous in opossum, and 71% homologous in cow. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title