Learn More
Description
This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein may play a role in bone development and fracture healing. Mutation in this gene, which is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7, may be associated with split-hand/split-foot malformation. [provided by RefSeq
Specifications
Specifications
| Antigen | DLX5 |
| Applications | ELISA, Immunohistochemistry (PFA fixed) |
| Classification | Monoclonal |
| Clone | 1B7 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant DLX5. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | DLX5 |
| Gene Accession No. | NM_005221 |
| Gene Symbols | DLX5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.