Learn More
distal-less homeobox 6, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human DLX6 protein.
Supplier: Abnova Corporation H00001750D01P
Description
This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. This family is comprised of at least 6 different members that encode proteins with roles in forebrain and craniofacial development. This gene is in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7. [provided by RefSeq
Sequence: MSHSQHSPYLQSYHNSSAAAQTRGDDTDQQKTTVIENGEIRFNGKGKKIRKPRTIYSSLQLQALNHRFQQTQYLALPERAELAASLGLTQTQVKIWFQNKRSKFKKLLKQGSNPHESDPLQGSAALSPRSPALPPVWDVSASAKGVSMPPNSYMPGYSHWYSSPHQDTMQRPQMMSpecifications
distal-less homeobox 6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human DLX6 protein. | |
DLX6 | |
MGC125282/MGC125283/MGC125284/MGC125285 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC069363.1 | |
DLX6 | |
DLX6 (AAH69363.1, 1 a.a. ∼ 175 a.a) full-length human protein. | |
100 μg | |
Neurobiology | |
1750 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.