Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Mouse anti-All, Clone: TNT-3, MilliporeSigma™

Catalog No. MAB3868MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3868MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3868MI Supplier MilliporeSigma Supplier No. MAB3868
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect DNA using this Anti-DNA Antibody, single stranded validated for use in ELISA, flow cytometry, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TNT-3
Conjugate Unconjugated
Formulation Protein A-purified murine IgG3 in PBS, carrier-free.
Host Species Mouse
Immunogen Human Raji cell nuclei
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.