Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-DNA-PK Clone: 18-2,

Catalog No. NA57100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA57100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA57100UG Supplier MilliporeSigma™ Supplier No. NA57100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects DNA-PK Clone: 18-2 in Human samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Immuno Blotting

DNA Dependent Protein Kinase, DNA-PK, is a nuclear serine/threonine protein kinase composed of two subunits, a ∽470kDa catalytic subunit, DNA-PKcs, and the Ku autoantigen, the heterodimeric DNA binding protein p70/p80. DNA-PK has been shown to phosphorylate a number of key proteins in vitro including p53, Sp1, myc, RPA, and RNA Pol I. DNA-PK activity requires association of DNA-PKcs with Ku, DNA binding, and the presence of DNA ends. Mutations in DNA-PKcs contribute to murine SCID phenotypes which leads to inefficient V(D)J recombination in developing T & B lymphocytes. Loss of DNA-PK activity in fibroblasts or lymphocytes also confers on cells hypersensitivity to killing by ionizing radiation. These two phenotypes implicate DNA-PK in the repair of DNA double strand breaks in all cells and V(D)J recombination in lymphocytes. The 4127 amino acid DNA-PKcs protein shares a high degree of similarity with the ATM family of kinases, including ATM, ATR, and FRAP in humans and MEC1 and TEL1 in yeast. The catalytic domain of DNA-PKcs also shares homology with the PI3K family of lipid kinases and DNA-PK is inhibitable by wortmanin, a lipid kinase inhibitor. While the exact role for DNA-PK in cell cycle regulation is unclear, the protein may be important for suppression of apoptosis since DNA-PKcs is targeted for proteolysis during apoptosis, which leads to loss of catalytic activity. DNA-PK can also be inactivated through phosphorylation, possibly by c-abl and by autophosphorylation. DNA-PKcs is expressed in all cells and a DNA-PK activity is observed in all higher eukaryotes examined. The protein level of DNA-PKcs in human cells is very high, 50 to 100x that observed in murine cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA-PK
Applications Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 18-2
Conjugate Unconjugated
Formulation In 10mM PBS, 0.2% BSA, pH 7.4.
Host Species Mouse
Immunogen DNA-PK purified from HeLa cells
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽470kDa DNA-PK protein in A431, HeLa, and HL60 cells, normal tonsil, normal colon, and breast carcinoma tissue.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.