Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase δ Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610972
Description
Errors in DNA sequence result from environmental factors or are committed by DNA polymerases during replication. If unchecked, these errors might accumulate genetic damage such that the cell could no longer function. Thus, DNA repair processes involve mechanisms for the excision of damaged sequences and the resynthesis and ligation of the proper sequence. In mammalian cells, this proofreading function rests with DNA polymerase (pol) δ, a heterodimer of a 50kDa subunit, which stimulates pol δ activity in the presence of PCNA (proliferating cell nuclear antigen) and a 125kDa catalytic subunit. The catalytic subunit has 3' to 5' exonuclease activity which distinguishes pol δ from pol α; and pol β;. Pol δ is also central to DNA replication where it functions in leading strand synthesis at the replication fork. The catalytic subunit is phosphorylated by G1 cyclin-dependent kinase-cyclin complexes and, via its N-terminal 249 amino acids, interacts with cdk2. However, phosphorylation has little or no effect on theactivity of pol δ. Thus, DNA polymerase δ is essential for DNA replication and is unique in its ability to replace damaged sequences through the process of DNA excision repair.
Specifications
DNA Polymerase δ | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human DNA Polymerase δ aa. 60-261 | |
50 μg | |
Cell Biology | |
Human, Rat | |
IgG1 |
Western Blot | |
22 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction