Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-DNA polymerase delta p125 Clone: 11E10.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABE967
Description
Specifically detects DNA polymerase delta p125 Clone: 11E10.1 in Human samples, and it is validated for Western Blotting
DNA polymerase delta catalytic subunit (EC 2.7.7.7; UniProt P28340; also known as CDC2 homolog, DNA polymerase subunit delta p125) is encoded by the POLD1 (also known as CDC2, MDPL, POLD, CRCS10) gene (Gene ID 5424) in human. DNA replication in eukaryotes involves multiple enzymes including the B family polymerases Pol α, Pol δ, and Pol ?. While Pol α initiates DNA synthesis, Pol δ and Pol ? replace Pol α after primer extension and perform the bulk of DNA replication. Pol δ and Pol ? are heterotetramers in higher eukaryotes comprised of POLD1 and POLE as the respective catalytic subunit, as well as accessory subunits (POLD2/3/4 and POLE2/3/4) that interact with cofactors such as Proliferating Cell Nuclear Antigen (PCNA). Both POLD1 and POLE contain an exonuclease domain (a.a. 268-471 of POLE and 304-517 of POLD1) that proof-reading capability during DNA replication. In addition to DNA synthesis, Pol ? and Pol δ are believed to participate in chromosomal DNA repairs, including nucleotide excision repair (NER), ismatch repair (MMR) and repair of double strand breaks (DSBR). Germline exonuclease domain mutations (EDMs) in POLD1 and POLE cause polymerase proofreading-associated polyposis (PPAP). Tumors with EDMs exhibits dramatically increased base substitutions.
Specifications
DNA polymerase delta p125 | |
Monoclonal | |
Unconjugated | |
P28340 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
11E10.1 | |
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
POLD1, POLD | |
GST-tagged recombinant protein corresponding to the N-terminus of human DNA polymerase delta p125. | |
100 μg | |
Epigenetics, Nuclear Function | |
NP_001243778 | |
2°C to 8°C for one year from date of shipment | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction