Learn More
DnaJ (Hsp40) homolog, subfamily A, member 3, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
DNAJA3 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins. For background information on the DNAJ family, see MIM 608375.[supplied by OMIM
Specifications
Specifications
| Antigen | DnaJ (Hsp40) homolog, subfamily A, member 3 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human DNAJA3 protein. |
| Formulation | No additive |
| Gene | DNAJA3 |
| Gene Accession No. | NM_005147 |
| Gene Alias | FLJ45758/TID1/hTid-1 |
| Gene Symbols | DNAJA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.