Learn More
DnaJ (Hsp40) homolog, subfamily B, member 6 (A01), Mouse anti-Human, Rat, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the 'J-domain' and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq]
Specifications
Specifications
| Antigen | DnaJ (Hsp40) homolog, subfamily B, member 6 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length recombinant DNAJB6. |
| Formulation | 50% glycerol |
| Gene | DNAJB6 |
| Gene Accession No. | BC000177 |
| Gene Alias | DJ4/DKFZp566D0824/DnaJ/FLJ42837/HHDJ1/HSJ-2/HSJ2/MGC1152/MGC117297/MRJ/MSJ-1 |
| Gene Symbols | DNAJB6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.