Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC25-GNG10, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206351 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-351 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-351 Supplier Abnova Corporation Supplier No. H00552891K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human DNAJC25-GNG10 peptide using ARM Technology.

This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen DNAJC25-GNG10
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description DNAJC25-GNG10 readthrough transcript
Formulation In 1X PBS, pH 7.4
Gene DNAJC25-GNG10
Gene Accession No. DNAJC25-GNG10
Gene Symbols DNAJC25-GNG10
Host Species Rabbit
Immunogen A synthetic peptide of human DNAJC25-GNG10
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552891
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.