Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DnaJ (Hsp40) homolog, subfamily C, member 7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018338 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-338 Supplier Abnova Corporation Supplier No. H00007266D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human DNAJC7 protein.

DNAJC7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM

Sequence: MAATEPELLDDQEAKREAETFKEQGNAYYAKKDYNEAYNYYTKAIDMCPKNASYYGNRAATLMMLGRFREALGDAQQSVRLDDSFVRGHLREGKCHLSLGNAMAACRSFQRALELDHKNAQAQQEFKNANAVMEYEKIAETDFEKRDFRKVVFCMDRALEFAPACHRFKILKAECLAMLGRYPEAQSVASDILRMDSTNADALYVRGLCLYYEDCIEKAVQFFVQALRMAPDHEKACIACRNAKALKAKKEDGNKAFKEGNYKLAYELYTEALGIDPNNIKTNAKLYCNRGTVNSKLRKLDDAIEDCTNAVKLDDTYIKAYLRRAQCYMDTEQYEEAVRDYEKVYQTEKTKEHKQLLKNAQLELKKSKRKDYYKILGVDKNASEDEIKKAYRKRALMHHPDRHSGASAEVQKEEEKKFKEVGEAFTILSDPKKKTRYDSGQDLDEEGMNMGDFDPNNIFKAFFGGPGGFSFEASGPGNFFFQFG

Specifications

Antigen DnaJ (Hsp40) homolog, subfamily C, member 7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human DNAJC7 protein.
Formulation PBS with no preservative; pH 7.4
Gene DNAJC7
Gene Accession No. NM_003315
Gene Alias DANJC7/DJ11/TPR2/TTC2
Gene Symbols DNAJC7
Host Species Rabbit
Immunogen DNAJC7 (NP_003306.1, 1 a.a. ∼ 484 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7266
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.