Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dynein, axonemal, light intermediate chain 1, Mouse, Clone: 2H3, Abnova™

Catalog No. 89001496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-496 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-496 Supplier Abnova Corporation Supplier No. H00007802M04A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DNALI1.

This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects. [provided by RefSeq

Sequence: MIPPADSLLKYDTPVLVSRNTEKRSPKARLLKVSPQQPGPSGSAPQPPKTKLPSTPCVPDPTKQAEEILNAILPPREWVEDTQLWIQQVSSTPSTRMDVVHLQEQLDLKLQQRQARETGICPVRRELYSQCFDELIREVTINCAERGLLLLRVRDEIRMTIAAYQTLYESSVAFGMRKALQAEQGKSDMERKIAELETEKRDLERQVNEQKAKCEATEKRESERRQVEEKKHNEEIQFLKRTNQQLKAQLEGIIAPKK

Specifications

Antigen dynein, axonemal, light intermediate chain 1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DNALI1.
Formulation ascites with no preservative
Gene DNALI1
Gene Accession No. BC046117
Gene Alias P28/dJ423B22.5/hp28
Gene Symbols DNALI1
Host Species Mouse
Immunogen DNALI1 (AAH46117, 1 a.a. ∼ 258 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7802
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.